Key Documents
SYNF234020C
Synthware chromatography column with vacuum connection and fritted disk
joint: ST/NS 24/40, O.D. × L 40 mm × 200 mm, Disc porosity: coarse
Synonym(s):
Synthware F234020C, glass chromatography column
About This Item
UNSPSC Code:
23151816
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F234020C
O.D. × L
40 mm × 200 mm
suitability
suitable for chromatography
General description
Short column with lower 24/40 joint and hose connection for vacuum. All Synthware columns are heavy wall designed to increase safety.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)