SLW1130/08D
Pyrex® Erlenmeyer flask, narrow neck, with printed trace code
capacity 125 mL
Synonym(s):
Pyrex, SciLabware 1130/08D
About This Item
Recommended Products
material
borosilicate glass
flat bottom
Joint
Joints neck (28)
sterility
non-sterile
feature
graduated
without cap
printed trace code
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1130/08D
H
117 mm
body O.D.
69 mm
capacity
125 mL
neck I.D.
28 mm
fitting
narrow neck
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with DIN ISO 1773
- Erlenmeyer pattern
- Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
- Graduated to full capacity
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service