DWK209425
Wheaton® Leak Resistant Bottle
capacity 30 mL, low-density polyethylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 33 mm × 69 mm, 28-410
Synonym(s):
209425
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
low-density polyethylene bottle
natural bottle
wide-mouth bottle
packaging
pkg of 72 ea
manufacturer/tradename
WHEATON® 209425
bottle diam. × H
33 mm × 69 mm
capacity
30 mL
Related Categories
Features and Benefits
Superior double seal closure for leak resistance
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)