Key Documents
BR708470
BRAND® EASYCAL micro-weighing container
Synonym(s):
Accessories, Calibration Software, EASYCAL
About This Item
UNSPSC Code:
41123300
NACRES:
NB.71
packaging
pack of 1 ea
manufacturer/tradename
BRAND 708470
General description
Pipettes < 50 μL can be surprisingly easy to calibrate using EASYCAL software and the new micro-weighing container. Includes 10 filters and 3 cover caps. For use with EASYCAL Calibration software (CD-ROM) .
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
EASYCAL is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)