Skip to Content
Merck
All Photos(3)

Key Documents

762695

Sigma-Aldrich

1H-Indene-2-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9BO2
CAS Number:
Molecular Weight:
159.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

57-77 °C

storage temp.

2-8°C

SMILES string

OB(O)C1=Cc2ccccc2C1

InChI

1S/C9H9BO2/c11-10(12)9-5-7-3-1-2-4-8(7)6-9/h1-5,11-12H,6H2

InChI key

VBMIZCKXRRAXBK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service