Skip to Content
Merck
All Photos(1)

Key Documents

297690

Sigma-Aldrich

2-Bromo-4,6-difluoroaniline

97%

Synonym(s):

2-Bromo-4,6-difluorobenzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(F)2NH2
CAS Number:
Molecular Weight:
208.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

41-42 °C (lit.)

functional group

bromo

SMILES string

Nc1c(F)cc(F)cc1Br

InChI

1S/C6H4BrF2N/c7-4-1-3(8)2-5(9)6(4)10/h1-2H,10H2

InChI key

WUJKFVGKLTWVSQ-UHFFFAOYSA-N

Application

2-Bromo-4,6-difluoroaniline may be used in chemical synthesis.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service