249580
9-Fluorenone-4-carbonyl chloride
97%
Synonym(s):
9-Oxo-4-fluorenecarbonyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H7ClO2
CAS Number:
Molecular Weight:
242.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
impurities
<2% dichloromethane
mp
139-141 °C (lit.)
storage temp.
2-8°C
SMILES string
ClC(=O)c1cccc2C(=O)c3ccccc3-c12
InChI
1S/C14H7ClO2/c15-14(17)11-7-3-6-10-12(11)8-4-1-2-5-9(8)13(10)16/h1-7H
InChI key
XKCGWCQMEUASJF-UHFFFAOYSA-N
General description
9-Fluorenone-4-carbonyl chloride is a fluorescent electrophilic reagent.
Application
9-Fluorenone-4-carbonyl chloride was used to functionalize amino acids in alkaline medium before their enantioresolution by HPLC. It was also used in the synthesis of N-(2-hydroxyethyl)-9-oxo-9H-fluorene-4 carboxamide.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
9-Fluorenon-4-carboxamides: synthesis, conformational analysis, anti-HSV-2, and immunomodulatory evaluation. Note II.
Alcaro S, et al.
ARKIVOC (Gainesville, FL, United States), 334, 348-348 (2004)
T-J Hsien et al.
Amino acids, 33(1), 97-104 (2006-10-28)
A fluorescent electrophilic reagent, 9-fluorenone-4-carbonyl chloride (FCC), is chosen to functionalize amino acids in alkaline medium before their HPLC resolution. FCC reacts with both primary and secondary amino acids to produce stable and highly fluorescent derivatives suitable for sensitive and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service