Merck
  • Development of acetophenone ligands as potential neuroimaging agents for cholinesterases.
Development of acetophenone ligands as potential neuroimaging agents for cholinesterases.
Bioorganic & medicinal chemistry (2016-09-18)
Courtney T Jollymore-Hughes, Ian R Pottie, Earl Martin, Terrone L Rosenberry, Sultan Darvesh
ABSTRACT
Association of cholinesterase with β-amyloid plaques and tau neurofibrillary tangles in Alzheimer's disease offers an opportunity to detect disease pathology during life. Achieving this requires development of radiolabelled cholinesterase ligands with high enzyme affinity. Various fluorinated acetophenone derivatives bind to acetylcholinesterase with high affinity, including 2,2,2-trifluoro-1-(3-dimethylaminophenyl)ethanone (1) and 1-(3-tert-butylphenyl)-2,2,2-trifluoroethanone (2). Such compounds also offer potential for incorporation of radioactive fluorine (
Sigma-Aldrich
Chlorodifluoroacetic anhydride, 98%
Sigma-Aldrich
N-Methoxy-N-methyl-2,2,2-trifluoroacetamide, 98%
Sigma-Aldrich
3-Bromo-N,N-dimethylaniline, 97%