Skip to Content
Merck

Skip To

1354207

USP

Isradipine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(4-Benzofurazanyl)-1,-4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid methyl 1-methylethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H21N3O5
CAS Number:
Molecular Weight:
371.39
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

isradipine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2cccc3nonc23)C(=O)OC(C)C

InChI

1S/C19H21N3O5/c1-9(2)26-19(24)15-11(4)20-10(3)14(18(23)25-5)16(15)12-7-6-8-13-17(12)22-27-21-13/h6-9,16,20H,1-5H3

InChI key

HMJIYCCIJYRONP-UHFFFAOYSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isradipine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Isradipine Capsules
  • Isradipine Compounded Oral Suspension

Biochem/physiol Actions

L-type calcium channel blocker (also referred to as dihydropyridine-type calcium channel blocker); antihypertensive.
L-type calcium channel blocker; antihypertensive.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y000036614632245.08507
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

USP

manufacturer/tradename

Calbiochem®

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Gene Information

-

API family

isradipine

API family

isradipine

API family

nicardipine

API family

-


Still not finding the right product?

Explore all of our products under Isradipine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1354207-200MG04061833851098

Questions

Reviews

No rating value

Active Filters