Skip to Content
Merck
All Photos(1)

Documents

UN10-1KT

Supelco

Fatty Acids Unsaturated Kit

(individualy packaged in quantities indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 1 kit

concentration

(individualy packaged in quantities indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

carboxylic acid

shipped in

dry ice

storage temp.

−20°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Arachidonic acid 100 mg
    cis-4,7,10,13,16,19-Docosahexaenoic acid 100 mg
    Elaidic acid 100 mg
    Erucic acid 100 mg
    Linoleic acid 100 mg
    Linolenic acid 100 mg
    Nervonic acid 100 mg
    Oleic acid 100 mg
    Palmitoleic acid 100 mg
    Petroselinic acid 100 mg
    See All (10)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhavna G Gordhan et al.
Scientific reports, 11(1), 6493-6493 (2021-03-24)
Rapid detection of tuberculosis (TB) infection is paramount to curb further transmission. The gold standard for this remains mycobacterial culture, however emerging evidence confirms the presence of differentially culturable tubercle bacteria (DCTB) in clinical specimens. These bacteria do not grow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service