PHR9219
2-Hydroxy-2,2-di(2-thienyl)acetic acid
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Tiotropium impurity A (PhEur), α-Hydroxy-α-2-thienyl-2-thiopheneacetic acid, 2-Hydroxy-2,2-di(thiophen-2-yl)acetic acid, Di-2-thienylglycolic acid
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
pharmaceutical small molecule
shipped in
wet ice
storage temp.
-10 to -25°C
SMILES string
[s]1c(ccc1)C(O)(c2[s]ccc2)C(=O)O
InChI
1S/C10H8O3S2/c11-9(12)10(13,7-3-1-5-14-7)8-4-2-6-15-8/h1-6,13H,(H,11,12)
InChI key
FVEJUHUCFCAYRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Application
Recommended products
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service