Z744122
Freezer box
W × L 3 in. × 3 in., without dividers
Synonym(s):
freezer container, freezer storage, storage container
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
white
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech FRSB-5000
technique(s)
cryopreservation: suitable
W × L
3 in. × 3 in.
Looking for similar products? Visit Product Comparison Guide
General description
3" Freezer Box, Each
Application
Cryogenics
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service