Z719536
Sorenson™ low binding Ultra G microtips
maximum volume 10 μL, pkg of 960 tips QuickRack, non-sterile
All Photos(1)
About This Item
Recommended Products
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
graduations
packaging
case of 4800 ea 50 QuickRack of 96 tips
pack of 960 ea 10 QuickRack of 96 tips
pkg of 960 tips QuickRack
manufacturer/tradename
Sorenson 30470T
maximum volume
10 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for ultra-micro pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.