Skip to Content
Merck
All Photos(1)

Key Documents

Z652229

Hettich® MIKRO 120 centrifuge accessories

adapter for 1.5 ml tubes 11x28 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

description

for use with 1252-A, 1258-A, 2434, 2437, 1195-A, 1189-A, 1158-L, 1154-L, 1555, 1552, 1553, 1789-A, 4790-A, 4489-A, 1226-A, 1213-A, 2428

feature

Holds 1.5 mL tubes

packaging

pack of 6 ea

manufacturer/tradename

Hettich 2031

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service