Skip to Content
Merck

SML3047

Urolithin C

≥97% (HPLC), Insulin secretion activator, powder

Synonym(s):

3,8,9-Trihydroxy-6H-benzo[c]chromen-6-one, 3,8,9-Trihydroxy-6H-dibenzo[b,d]pyran-6-one, 3,8,9-Trihydroxyurolithin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H8O5
CAS Number:
Molecular Weight:
244.20
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Urolithin C, ≥97% (HPLC)

Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C2=CC(O)=C(O)C=C2C3=C(C=C(O)C=C3)O1

InChI key

HHXMEXZVPJFAIJ-UHFFFAOYSA-N

Biochem/physiol Actions

Urolithin C is chebulic ellagitannins metabolite that exerts antiproliferative and antioxidant activity. Urolithin C exhibit glucose dependent enhancement of insulin secretion in islet cells under normal or dysfunctional conditions. Apparently, it facilitates L-type Ca2+ channel opening and Ca2+ influx into pancreatic β-cells.
chebulic ellagitannins metabolite, exhibit glucose dependent enhancement of insulin secretion


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library