Skip to Content
Merck
All Photos(1)

Key Documents

SML3045

Sigma-Aldrich

XL177B

≥98% (HPLC)

Synonym(s):

(R)-N-(4-benzyl-5-(4-hydroxy-4-((7-(3-(4-methylpiperazin-1-yl)propanamido)-4-oxoquinazolin-3(4H)-yl)methyl)piperidin-1-yl)-5-oxopentyl)-9-chloro-5,6,7,8-tetrahydroacridine-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H57ClN8O5
CAS Number:
Molecular Weight:
861.47
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(CC2(O)CCN(C([C@@H](CC3=CC=CC=C3)CCCNC(C4=CC(N=C(CCCC5)C5=C6Cl)=C6C=C4)=O)=O)CC2)C=NC7=CC(NC(CCN8CCN(C)CC8)=O)=CC=C71

InChI key

UYJWYMWIVMCGQZ-PGUFJCEWSA-N

Biochem/physiol Actions

XL177B is a negative enantiomeric control for a cell penetrant, potent and highly selective covalent inhibitor of the deubiquitinating enzyme USP7 XL177A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan J Schauer et al.
Scientific reports, 10(1), 5324-5324 (2020-03-27)
Ubiquitin specific peptidase 7 (USP7) is a deubiquitinating enzyme (DUB) that removes ubiquitin tags from specific protein substrates in order to alter their degradation rate and sub-cellular localization. USP7 has been proposed as a therapeutic target in several cancers because
Jonathan W Bushman et al.
Cell chemical biology, 28(1), 78-87 (2020-10-03)
Deubiquitinating enzymes (DUBs) catalyze the removal of ubiquitin, thereby reversing the activity of E3 ubiquitin ligases and are central to the control of protein abundance and function. Despite the growing interest in DUBs as therapeutic targets, cellular functions for DUBs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service