Skip to Content
Merck
All Photos(1)

Key Documents

SAE0086

Sigma-Aldrich

Peptidyl Arginine Deiminase Type 4

human recombinant, expressed in E. coli, aqueous solution

Synonym(s):

Peptidyl Arginine Deiminase Type 4, PAD4, PADI4, Protein arginine iminohydrolase 4

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
3.5.3.15
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

specific activity

≥200 units/mg protein

mol wt

74 kDa

UniProt accession no.

application(s)

genomic analysis

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PADI4(23569)

General description

Peptidyl Arginine Deiminases (PADs) perform post-translational deiminations of proteins. PADs are calcium-dependent enzymes that catalyze the conversion of L-arginine residues to L-citrulline. The catalyzed reaction is as follows:

2+
Ca Protein--[L-arginine] + H2O ––––> Protein--[L-citrulline]

There are five mammalian PADs sub-types, which differ in substrate specificity and tissue distribution. PAD enzymes are highly homologous, with 50–60% sequence similarity.1 PADs play important roles in gene regulation by citrullination of arginine residues on histones H3, H2A, and H4. Overexpression of PADs has been found in conditions such as rheumatoid arthritis, Alzheimer′s disease, multiple sclerosis, lupus,Parkinson′s disease, and cancer.2,3

PAD4 is widely expressed in the immune system with expression pathology in various tumor and cancer cell lines.5 In cells, PAD4 can be found in the cytoplasm and nucleus. PAD4 activity has been correlated to the development and progress of Rheumatoid Arthritis (RA).2,5 A study in mice has indicated a mechanism for the development of RA by anticitrullin autoimmunity.6

This recombinant human PAD4 is expressed in Escherichia coli as a fusion protein. The fusion partner is removed by HRV3C protease digestion, followed by chromatographic purification.

Biochem/physiol Actions

Calcium is required for peptidylarginine deiminase activity in vitro.

Unit Definition

One unit will produce 1 μmole of N-a-benzoylcitrulline ethyl ester from BAEE per hour at 37 °C at pH 7.7.

Physical form

Supplied as an aqueous solution in 10 mM Tris (pH 7.5), 500 mM NaCl, 1 mM EDTA, and 1 mM DTT.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pekka Rappu et al.
Frontiers in oncology, 12, 1035188-1035188 (2022-12-20)
Protein arginine deiminases (PADs) are intracellular enzymes that may, especially in pathological conditions, also citrullinate extracellular substrates, including matrisome proteins such as structural proteins in extracellular matrix (ECM). PADs are abundantly expressed in human cancer cells. Citrullination of matrisome proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service