Skip to Content
Merck
All Photos(1)

Key Documents

PZ0381

Sigma-Aldrich

PF-06651385

≥98% (HPLC)

Synonym(s):

4-(3-Chloro-4-(trifluoromethoxy)benzyloxy)-N-(1,2,4-thiadiazol-5-yl)benzenesulfonamide, PF 06651385, PF06651385

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11ClF3N3O4S2
CAS Number:
Molecular Weight:
465.85
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=CC(COC2=CC=C(S(NC3=NC=NS3)(=O)=O)C=C2)=CC=C1OC(F)(F)F

InChI

1S/C16H11ClF3N3O4S2/c17-13-7-10(1-6-14(13)27-16(18,19)20)8-26-11-2-4-12(5-3-11)29(24,25)23-15-21-9-22-28-15/h1-7,9H,8H2,(H,21,22,23)

Biochem/physiol Actions

PF-06651385 is an Nav1.3 voltage-gated sodium channel modulator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service