GE80-1261-01
ExcelGel™precast gels for SDS-PAGE
12.5 gel concentration, pack of 6 ea
About This Item
Recommended Products
material
polyacrylamide
polyester film
form
solid
feature
wetted part: no
packaging
pack of 6 ea
manufacturer/tradename
Cytiva 80126101
parameter
15-40 kDa separation range
L × thickness
110 mm × 0.5 mm
color
Colorless
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
ExcelGel gradient gels are recommended for use with Immobiline DryStrip gels.
Features and Benefits
- Save time with ready-to-use, mechanically stable gels cast on plastic supports.
- Apply samples quickly and easily using preformed sample wells.
- Avoid making running buffer by using precast buffer strips.
Storage and Stability
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service