Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0963-02

Protein A Sepharose Cl-4B

Cytiva 17-0963-02, pack of 500 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

native protein A

packaging

pack of 500 mL

manufacturer/tradename

Cytiva 17-0963-02

storage condition

(20% Ehtanol)

matrix

4% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

2-10

working range

3-9

capacity

~40 mg binding capacity (histidine-tagged protein/ml medium when changed with Ni2+)(Protein- and metal-ion dependent.)

storage temp.

2-8°C

General description

Protein A Sepharose CL-4B is a classic, well documented medium for immunoglobin purification and fractionation, and is suitable for immunoprecipitation procedures.

Protein A binds to the Fc region of immunoglobulins through interactions with the heavy chain. The binding of protein A has been well documented for IgG from a variety of mammalian species and for some IgM and IgA as well. Protein A Sepharose CL-4B has been used as a powerful tool to isolate and purify classes, subclasses and fragments of immunoglobulins from biological fluids and from cell culture media. Since only the Fc region is involved in binding, the Fab region is available for binding antigen.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • Well documented for immunoglobulin purification and fractionation
  • Strong binding to Fc region of IgG
  • High capacity - 20 mg human IgG/mL.
  • Suited for immunoprecipitation procedures

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service