Skip to Content
Merck
All Photos(1)

Key Documents

EP5804771001

Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL

holds 2 tubes 7-15 mL, pack of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

feature

holds 2 tubes 7-15 mL

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5804771001

technique(s)

centrifugation: suitable

compatibility

for use with Rotor F-34-6-38

Looking for similar products? Visit Product Comparison Guide

General description

  • For 2 round-bottom tubes 7–15 mL.
  • For use with Rotor F-34-6-38.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service