Skip to Content
Merck
All Photos(1)

Key Documents

EHU089141

Sigma-Aldrich

MISSION® esiRNA

targeting human ATXN3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTGTGTGATTACAGCATAGGGTCCACTTTGGTAATGTGTCAAAGAGATGAGGAAATAAGACTTTTAGCGGTTTGCAAACAAAATGATGGGAAAGTGGAACAATGCGTCGGTTGTAGGACTAAATAATGATCTTCCAAATATTAGCCAAAGAGGCATTCAGCAATTAAAGACATTTAAAATAGTTTTCTAAATGTTTCTTTTTCTTTTTTGAGTGTGCAATATGTAACATGTCTAAAGTTAGGGCATTTTTCTTGGATCTTTTTGCAGACTAGCTAATTAGCTCTCGCCTCAGGCTTTTTCCATATAGTTTGTTTTCTTTTTCTGTCTTGTAGGTAAGTTGGCTCACATCATGTAATAGTGGCTTTCATTTCTTATTAACCAAATTAACCTTTCAGGAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingfeng Tu et al.
Nucleic acids research, 45(8), 4532-4549 (2017-02-10)
The Chk1 protein is essential for genome integrity maintenance and cell survival in eukaryotic cells. After prolonged replication stress, Chk1 can be targeted for proteasomal degradation to terminate checkpoint signaling after DNA repair finishes. To ensure proper activation of DNA
Avraham Ashkenazi et al.
Nature, 545(7652), 108-111 (2017-04-27)
Nine neurodegenerative diseases are caused by expanded polyglutamine (polyQ) tracts in different proteins, such as huntingtin in Huntington's disease and ataxin 3 in spinocerebellar ataxia type 3 (SCA3). Age at onset of disease decreases with increasing polyglutamine length in these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service