Key Documents
CLS432076
Corning® CoolCell Filler Vials, for use with CoolCell LX and CoolCell FTS30 containers
2 mL
Synonym(s):
cryovials
About This Item
UNSPSC Code:
41103025
NACRES:
NB.12
packaging
pkg of 6
manufacturer/tradename
Corning 432076
capacity
2 mL
General description
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. It is suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5ml LX containers.
Legal Information
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)