C-39427
Preadipocyte Growth Medium
SupplementPack containing all media supplements as individual vials, 1 Pack for 500 ml
Synonym(s):
Recommended with C-27411
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
liquid
packaging
pkg of 1 kit
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Preadipocyte Growth Medium SupplementPack (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-27411
To form a complete medium, it should be combined with the basal medium: C-27411
Application
Preadipocyte Growth Medium is a low-serum (5% V/V) medium developed for the undifferentiated expansion of human preadipocytes. The media are optimized for primary human cells, but can also be used for murine preadipocytes.Recommended for:Human White Preadipocytes (HWP)
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Preadipocyte/Adipocyte Media are subjected to comprehensive quality control tests using primary human white preadipocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested preadipocytes. In addition, the Preadipocyte Differentiation Medium/Adipocyte Nutrition Medium are tested for the capacity to induce the terminal differentiation of preadipocytes into mature adipocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.05 ml / mlEndothelial Cell Growth Supplement: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 10 ng / mlHydrocortisone: 1 µg / mlHeparin: 90 µg / ml
Storage and Stability
Store the SupplementPack at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
recommended
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service