C-20211
Keratinocyte Growth Medium 2
Basal Medium, 500 ml
Synonym(s):
Recommended with C-39016 or C-39011
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
aqueous solution
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
storage temp.
2-8°C
General description
Keratinocyte Basal Medium 2 (supplements sold separately)
To form a complete medium, it should be combined with the media supplements: C-39016 or C-39011
To form a complete medium, it should be combined with the media supplements: C-39016 or C-39011
Application
Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells. The medium is optimized for primary human cells but can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.Recommended for:Normal Human Epidermal Keratinocytes (NHEK), juvenile foreskinNormal Human Epidermal Keratinocytes (NHEK), adult donor
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Keratinocyte Growth Medium 2 are subjected to comprehensive quality control tests using primary human keratinocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested keratinocytes. Approved inhouse lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Storage and Stability
Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C
Subculture Routine
Click here for more information.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service