Skip to Content
Merck
All Photos(1)

Key Documents

A9227

Sigma-Aldrich

ABT-491 hydrochloride

≥98% (HPLC)

Synonym(s):

4-Ethynyl-N,N-dimethyl-3-[3-fluoro-4-[(2-methyl-1H-imidazo-[4,5-c]pyridin-1-yl)methyl]benzoyl]-1H-indole-1-carboxamide hydrochloride, A-137491

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22N5O2F · HCl
CAS Number:
Molecular Weight:
515.97
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to beige

mp

170-178 °C (lit.)

solubility

H2O: freely soluble 22 mg/mL

originator

Abbott

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)C(=O)n1cc(C(=O)c2ccc(Cn3c(C)nc4cnccc34)c(F)c2)c5c(cccc15)C#C

InChI

1S/C28H22FN5O2.ClH/c1-5-18-7-6-8-25-26(18)21(16-34(25)28(36)32(3)4)27(35)19-9-10-20(22(29)13-19)15-33-17(2)31-23-14-30-12-11-24(23)33;/h1,6-14,16H,15H2,2-4H3;1H

InChI key

AWRGBOKANQBIBM-UHFFFAOYSA-N

Biochem/physiol Actions

Highly potent, selective and orally active platelet-activating factor (PAF) receptor antagonist. Ki = 0.6 nM in human platelets.

Features and Benefits

This compound is featured on the PAF Receptor page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service