Skip to Content
Merck

19475

Aminoallyl-dUTP-Cy®5 triethylammonium salt solution

≥95.0% (HPLC)

Synonym(s):

5-(3-Aminoallyl)-2′-deoxyuridine 5′-triphosphate, labeled with Cy®5 triethylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H58N5O21P3S2 · xC6H15N
Molecular Weight:
1162.01 (free acid basis)
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (HPLC)

form

liquid

pH

7.0-8.0

storage temp.

−20°C

SMILES string

OP(OP(O)(OP(O)(OC[C@@H]1[C@@H](O)C[C@H](N2C(NC(C(/C=C/CNC(CCCCCN(C3=C(C/4(C)C)C=C(S(=O)(O)=O)C=C3)C4=C\C=C\C=C\C5=[N+](CC)C6=CC=C(S(=O)([O-])=O)C=C6C5(C)C)=O)=C2)=O)=O)O1)=O)=O)(O)=O.CCN(CC)CC

InChI

1S/C45H58N5O21P3S2.C6H15N/c1-6-48-34-20-18-30(75(62,63)64)24-32(34)44(2,3)38(48)15-9-7-10-16-39-45(4,5)33-25-31(76(65,66)67)19-21-35(33)49(39)23-12-8-11-17-40(52)46-22-13-14-29-27-50(43(54)47-42(29)53)41-26-36(51)37(69-41)28-68-73(58,59)71-74(60,61)70-72(55,56)57;1-4-7(5-2)6-3/h7,9-10,13-16,18-21,24-25,27,36-37,41,51H,6,8,11-12,17,22-23,26,28H2,1-5H3,(H7-,46,47,52,53,54,55,56,57,58,59,60,61,62,63,64,65,66,67);4-6H2,1-3H3/b14-13+;/t36-,37-,41-;/m1./s1

InChI key

YSKSFNTVPAYJLX-OHCXCEGRSA-N

Legal Information

Cy is a registered trademark of Cytiva


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library