Skip to Content
Merck
All Photos(1)

Key Documents

ST407

Sigma-Aldrich

SilaCOgen

≥95.0% (GC)

Synonym(s):

Methyldiphenylsilanecarboxylic acid, 1-Methyl-1,1-diphenylsilanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O2Si
CAS Number:
Molecular Weight:
242.35
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

reaction suitability

reaction type: C-C Bond Formation

InChI key

JGZJTJGZAGJOOE-UHFFFAOYSA-N

Application

SilaCOgen is a silanecarboxylic acid whose decomposition is used as a source of CO.

Learn more about CO releasing molecules from Prof. Skrydstrup

SilaCOgen can be used with COware Platform

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service