Skip to Content
Merck
All Photos(1)

Key Documents

PHL80180

Urushiol (15:1)

phyproof® Reference Substance

Synonym(s):

3-(8Z-Pentadecenyl) 1,2-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥85.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

Oc1c(cccc1CCCCCCC\C=C/CCCCCC)O

InChI

1S/C21H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-17-15-18-20(22)21(19)23/h7-8,15,17-18,22-23H,2-6,9-14,16H2,1H3/b8-7-

InChI key

GWOCLAPCXDOJRL-FPLPWBNLSA-N

Related Categories

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Extraction and Analysis of Urushiol 15:1: A method utilizing vortex-blending matrix solid-phase dispersion coupled with UPLC-Q-TOF/MS has been developed to extract and analyze urushiol from the bark of Toxicodendron vernicifluum. This approach offers a refined technique for studying urushiol compounds and their allergenic effects, relevant to urushiol-induced contact dermatitis research (Chen et al., 2024).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service