Skip to Content
Merck
All Photos(1)

Key Documents

63367

Millipore

Dextrose Disks

suitable for microbiology, Sterile filter paper discs impregnated with dextrose

Synonym(s):

Carbohydrate discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

Quality Level

sterility

sterile

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 10 × 25 discs

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
Neisseria spp.
Proteus spp.
Salmonella spp.
Serratia spp.
Shigella spp.
Streptococcus spp.
bacteria

General description

Dextrose Discs are used to differentiate bacteria based on the dextrose fermentation ability. Microorganisms can only ferment certain carbohydrates, depending on the enzymes they possess to break down carbohydrates. Fermentation can be detected by gas production (CO2) in liquid media and/or color change of pH caused by acid production. The dextrose discs are aseptically added to any sugar-free basal media to test the ability of the organism to hydrolyze dextrose, a simple monosaccharide, as the sole source of carbon and energy during fermentation. Dextrose is typically fermented under acid production by many Enterobacter and coliform bacteria.

Application

Dextrose Discs are recommended to detect dextrose fermenting bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service