Skip to Content
Merck
All Photos(1)

Key Documents

05284

Supelco

Triethylene glycol dimethyl ether

analytical standard

Synonym(s):

1,2-Bis(2-methoxyethoxy)ethane, Dimethyltriglycol, NSC 66400, Triglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)3CH3
CAS Number:
Molecular Weight:
178.23
Beilstein:
1700630
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>4.7 (vs air)

Assay

≥98% (GC)

autoignition temp.

375 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

APHA: ≤20

refractive index

n20/D 1.423 (lit.)

bp

216 °C (lit.)

mp

−45 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

COCCOCCOCCOC

InChI

1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3

InChI key

YFNKIDBQEZZDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylene glycol dimethyl ether may be used as an analytical standard for the quantification of the analyte in water samples using in vitro bioassays and chemical screening technique.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability of superoxide radicals in glyme solvents for non-aqueous Li?O 2 battery electrolytes
Schwenke UK
Physical Chemistry Chemical Physics, 15(28), 11830-11839 (2013)
Sample preparation for combined chemical analysis and in vitro bioassay application in water quality assessment
Kolkman A, et al.
Environmental Toxicology and Pharmacology, 36(3), 1291-1303 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service