Skip to Content
Merck
All Photos(1)

Key Documents

00900585

Berberine chloride

primary reference standard

Synonym(s):

Berberine chloride form, Natural Yellow 18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO4
CAS Number:
Molecular Weight:
371.81
Colour Index Number:
75160
Beilstein:
3836585
MDL number:
UNSPSC Code:
12352210
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

SMILES string

COC1=C(C=[N+](CCC2=C3C=C4C(OCO4)=C2)C3=C5)C5=CC=C1OC.[Cl-]

InChI

1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1

InChI key

VKJGBAJNNALVAV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Fluorescent stain for heparin in mast cells

Biochem/physiol Actions

An alkaloid with weak antibiotic properties. Substrate for MDR efflux pumps. Antimicrobial activities of berberine is potentiated by the MDR inhibitor 5´-methoxyhydnocarpin (5´-MHC). Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service