Select a Size
About This Item
Product Name
SNAP id® 2.0 Midi Blot Holding Frame (single pack), The new Midi Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping & waste management.
Quality Segment
manufacturer/tradename
SNAP id®
technique(s)
western blot: suitable
compatibility
for use with Commercially available blocking reagents, for use with Luminata Western HRP Substrates, for use with Nitrocellulose, for use with PVDF (Immobilon membranes), for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with commercially available detection reagents
detection method
chemiluminescent, colorimetric, fluorometric
shipped in
ambient
General description
Application
Legal Information
Disclaimer
Still not finding the right product?
Explore all of our products under SNAP id® 2.0 Midi Blot Holding Frame (single pack)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.