Skip to Content
Merck

PCC030C45E

Millipore

Pellicon® Capsule with Ultracel® Membrane

30 kDa, C Screen, 4.5 m2 manifold extender with AseptiQuik® L connectors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903

material

EPDM rubber seal
PPO/PS blend (Core)
PPO/PS blend housing
TPE seal
composite regenerated cellulose (CRC) membrane
epoxy adhesive
polyester screen
polypropylene screen
polyurethane adhesive

Quality Level

sterility

sterile; γ-irradiated

product line

Pellicon®

storage condition

room temperature (15-30°C)

parameter

4-8 L/min-m2 flow rate
50 psi pressure (3.5 bar) at 4-30 °C (Forward Transmembrane Pressure (TMP))
60 psi max. pressure (4.1 bar) at 4-30 °C (Maximum Inlet Pressure)

technique(s)

ultrafiltration: suitable

H

46.9 cm (18.5 in.)

L

60.2 cm (23.7 in.)

W

48.2 cm (19.0 in.)

filtration area

4.5 m2

screen size

, Type C screen (coarse screen)

volume

1857 mL , in feed channel
2403 mL , in permeate channel

impurities

≤5 ppm TOC ( in retentate effluent, after a WFI flush of 20 L/m²)

matrix

Ultracel®

pore size

30 kDa

pH range

2-13

fitting

inlet/outlet connection (Feed: AseptiQuik® L Connector)
inlet/outlet connection (Permeate: AseptiQuik® L Connector)
inlet/outlet connection (Retentate: AseptiQuik® L Connector)

General description

The Pellicon® Capsule is the first of its kind -- a true single-use TFF device that comes ready to process in minutes. Self-contained, it employs an easy-to-use, holderless format and is supplied sterilized by irradiation.

Application

Concentration, Diafiltration, Buffer exchange, Protein purification

Features and Benefits

Plug ′n play, holderless design - easy to install, safe to remove
Gamma Sterilized and preseravtive-free - ready to process in minutes
True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families

Packaging

The manifold assembly is inserted into a polyethylene foam frame and then double bagged and vacuum sealed. The double-bagged assembly is placed within a second foam frame and individually boxed (qty 1 per box). Packaged assemblies are palletized.

Other Notes

The Emprove® Program complements the product portfolio through
Comprehensive documentation to support qualification, risk assessment and process optimization needs.
Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
Convenient 24/7 access to up-to-date product information.

Legal Information

AseptiQuik is a registered trademark of Colder Products Company
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service