Skip to Content
Merck
All Photos(1)

Key Documents

MXPESP5VT

Millipore

Fluoroelastomer O-ring + Plug Seal

Accessory for On-line sampling and filtration of aqueous liquids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.24

material

fluoroelastomer

Quality Level

packaging

pkg of 5 numbers

manufacturer/tradename

MicropreSURE®

input

sample type wine
sample type dairy
sample type cider
sample type pharmaceutical(s)
sample type beer
sample type food(s)
sample type sport drink
sample type bottled water
sample type soft drink
sample type beverage(s)

application(s)

bioburden testing
food and beverages
pharmaceutical

shipped in

ambient

General description

Fluoroelastomer O-ring + Plug Seal is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The fluoroelastomer O-ring + plug seal aids in the sealed and leak-free flow of the sample.

Application

Fluoroelastomer O-ring + plug seal is used for in-process sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service