MS0PENR01
MSOpener Vacuum Manifold
For accessing the membrane used on-line sampling and filtration of aqueous liquids
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.24
Recommended Products
Quality Level
packaging
pkg of 1 ea
manufacturer/tradename
MicropreSURE®
input
sample type food(s)
sample type beverage(s)
application(s)
bioburden testing
clinical
environmental
food and beverages
industrial qc
shipped in
ambient
General description
MSOpener Vacuum Manifold is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The MSOpener vacuum manifold is used to access the membrane post-filtration by connecting to a vacuum source and placing the filtration sampler on the MSOpener.
Application
MSOpener vacuum manifold is used in on-line sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.
Legal Information
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service