HAWGPDMCB
Total Coliform and E. coli kit
A Kit for simultaneous detection of coliform, E.coli using m-coli Blue 24 with S-Pak® membranes and sterile petri dish with petri-pad
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NA.74
Recommended Products
storage condition
protect from light
pore size
0.45 μm pore size
application(s)
cosmetics
environmental
food and beverages
water monitoring
shipped in
wet ice
storage temp.
2-8°C
General description
Total Coliform and E.coli Kit contains:
Petri-Pad Petri dish is 47mm sterilized Petri dishes for use with solid agar medium. Pre-sterilized disposable 47mm dishes preloaded with sterilized absorbent pads minimize contamination.
S-Pak® Filters are made of Mixed cellulose Esters (MCE) with 0.22 μm pore size, white gridded, 47mm diameter.
m-ColiBlue24 Broth allows for the simultaneous detection of total coliform bacteria and Escherichia coli (E. coli) within 24 hours. An enzymatic indicator in the medium causes non-fecal total coliform colonies grown on the m-ColiBlue24 medium to be red, while the E. coli (fecal coliform) colonies are blue. The selectivity of the enzymatic indicator eliminates the need for confirmation tests.
Petri-Pad Petri dish is 47mm sterilized Petri dishes for use with solid agar medium. Pre-sterilized disposable 47mm dishes preloaded with sterilized absorbent pads minimize contamination.
S-Pak® Filters are made of Mixed cellulose Esters (MCE) with 0.22 μm pore size, white gridded, 47mm diameter.
m-ColiBlue24 Broth allows for the simultaneous detection of total coliform bacteria and Escherichia coli (E. coli) within 24 hours. An enzymatic indicator in the medium causes non-fecal total coliform colonies grown on the m-ColiBlue24 medium to be red, while the E. coli (fecal coliform) colonies are blue. The selectivity of the enzymatic indicator eliminates the need for confirmation tests.
Application
Total Coliform and E.coli Kit with S-Pak® membranes and sterile petri dish with petri-pad are used in detection of total coliform bacteria in Beer, Bottled Water, Cider, Sport Drink, Wine etc.
Components
600 S-pak filters (HAWG047S6)
150 Petri pad (MSP010063)
150 m-ColiBlue ampoules (M00PMCB24)
150 Petri pad (MSP010063)
150 m-ColiBlue ampoules (M00PMCB24)
Legal Information
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service