Skip to Content
Merck
All Photos(1)

Key Documents

CGW372S48

Millipore

Polysep II 1.0/0.2 µm nominal, Cartridge

cartridge nominal length 20 in. (50 cm), filter diam. 2.8 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

material

borosilicate glass fiber (BGF) membrane
mixed cellulose esters (MCE) membrane
polypropylene device
polypropylene support (spun bonded)
silicone seal

Quality Level

reg. compliance

meets FDA 21 CFR 177–182 (all component materials)

sterility

non-sterile

Sterilization Compatibility

autoclavable compatible
steam-in-place compatible

product line

Polysep

feature

hydrophilic

manufacturer/tradename

Polysep II

parameter

3.5 bar max. differential pressure (50 psid) at 25 °C

technique(s)

prefiltration: suitable

L

20 in.

W

2.7 in.

cartridge nominal length

20 in. (50 cm)

device size

20 in.

filter diam.

2.8 in.

filter filtration area

0.92 m2

impurities

≤0.5 EU/mL bacterial endotoxins (LAL test, aqueous extraction)
<0.5 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)

matrix

Polysep II

pore size

1.0/0.2 μm nominal pore size
1.0/0.2 μm pore size

cartridge code

Code 7

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Cartridge
Cartridge Type: Liquid
Filter Grade: Prefilter

Application

mAb & r-protein intermediates; Plasma; Vaccines & viral vectors; SVPs LVPs; Cell Culture; Media Filtration; Colloid Removal; Lipid Removal

Features and Benefits

Format: Double Layer

Packaging

Double Easy-Open bag

Preparation Note

Sanitization: 30 min with hot water at 80 °C
Sterilization Method: 10 autoclave cycles or SIP at 121 °C for 30 min
This product was manufactured with a Millipore® media combination which meets the criteria for a "non-fiber releasing" filter as defined in 21 CFR 210.3 (b) (6).

Analysis Note

After autoclaving and a 24 hour soak in ASTM® Type 1 reagent grade water at controlled room temperature, extractables level was ≤140 mg per 10-inch element after a 5000 mL flush.
Cartridges meet the requirements of the USP Oxidizable Substances Test after a water flush of 5000 mL per 10 inch Element

Other Notes

Directions for Use:
  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer first page of Visual Inspection Guide and introduction section of Mixed cellulose esters Tech Brief
  • Storage Statement: Please refer Mixed cellulose esters Tech Brief
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations

The product is manufuactured in a facility with a Quality Management System approved by an accredited registering body to an ISO 9001 Quality Systems Standard.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Polysep is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service