Skip to Content
Merck
All Photos(1)

Key Documents

8.20290

Sigma-Aldrich

2-Chloro-N,N-dimethylacetamide

for synthesis

Synonym(s):

2-Chloro-N,N-dimethylacetamide, N,N-Dimethyl-2-chloroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CON(CH3)2
CAS Number:
Molecular Weight:
121.57
MDL number:
UNSPSC Code:
12352111
EC Index Number:
220-224-4
NACRES:
NA.22

Quality Level

Assay

≥96.5% (GC)

form

liquid

bp

98-100 °C/15 hPa

density

1.18 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCC(=O)N(C)C

InChI

1S/C4H8ClNO/c1-6(2)4(7)3-5/h3H2,1-2H3

InChI key

XBPPLECAZBTMMK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Density (d 20 °C/ 4 °C): 1.180 - 1.186
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service