8.14439
2-Propylvaleric acid
for synthesis
Synonym(s):
2-Propylvaleric acid, Dipropylacetic acid, Valproic acid, 2-Propylpentanoic acid
About This Item
Recommended Products
vapor pressure
0.01 hPa ( 20 °C)
Quality Level
form
liquid
potency
670 mg/kg LD50, oral (Rat)
expl. lim.
1 % (v/v)
bp
220 °C/1013 hPa
transition temp
flash point 111 °C
density
0.905 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
OC(=O)C(CCC)CCC
InChI
1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)
InChI key
NIJJYAXOARWZEE-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1A - Skin Irrit. 2
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
231.8 °F - closed cup
Flash Point(C)
111 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service