Skip to Content
Merck
All Photos(1)

Key Documents

5.32628

Sigma-Aldrich

Vps34 Inhibitor, VPS34-IN1

VPS34-IN1

Synonym(s):

Vps34 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

SMILES string

Clc1nccc(c1)Nc2nc(ccn2)c3c(nc(nc3)NCC(O)(C)C)CC4CC4

InChI key

AWNXKZVIZARMME-UHFFFAOYSA-N

Packaging

Glass bottle

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service