Skip to Content
Merck
All Photos(1)

Documents

1.12285

Sigma-Aldrich

Niobium(V) chloride

98+

Synonym(s):

Niobium(V) chloride, Niobium pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl5Nb
CAS Number:
Molecular Weight:
270.17
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-059-8
NACRES:
NA.22

Quality Level

form

solid

potency

1400 mg/kg LD50, oral (Rat)

mp

205 °C

density

2.75 g/cm3

bulk density

1450 kg/m3

storage temp.

no temp limit

InChI

1S/5ClH.Nb/h5*1H;/q;;;;;+5/p-5

InChI key

YHBDIEWMOMLKOO-UHFFFAOYSA-I

Analysis Note

Identity: passes test
Assay (gravimetrically): ≥ 98.0 %

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service