Skip to Content
Merck
All Photos(1)

Key Documents

1.00594

Supelco

Aluminum Test, photometric

photometric, 0.02-0.50 mg/L (Al), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Aluminium Cell Test, photometric, 0.02-0.50 mg/L (Al), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

aluminium

measuring range

0.02-0.50 mg/L (Al)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric ( Chromazurol S )

storage temp.

15-25°C

General description

Method: photometric 0.02 - 0.50 mg/l Al Spectroquant®

The method is analogous to APHA 3500-Al B and DIN ISO 10566.


This Spectroquant Aluminium Cell Test allows the accurate quantification of the aluminium content in aqueous samples.


Method applied: In weakly acidic, acetate-buffered solution aluminium ions react with chromazurol S to form a blue-violet compound that is determined photometrically.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service