Skip to Content
Merck
All Photos(1)

Key Documents

999994P

Avanti

12:0 Diether PC

Avanti Research - A Croda Brand 999994P, powder

Synonym(s):

1,2-di-0-dodecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H68NO6P
CAS Number:
Molecular Weight:
593.86
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (999994P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999994P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCC)COCCCCCCCCCCCC)=O

InChI

1S/C32H68NO6P/c1-6-8-10-12-14-16-18-20-22-24-27-36-30-32(31-39-40(34,35)38-29-26-33(3,4)5)37-28-25-23-21-19-17-15-13-11-9-7-2/h32H,6-31H2,1-5H3/t32-/m1/s1

InChI key

MIOIPSRFEJKMJG-JGCGQSQUSA-N

General description

12:0 Diether PC (phosphocholine) is a glycerophospholipid having choline as the alcohol moiety, attached to the phosphate group. It has two dodecane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds.

Application

12:0 Diether PC or 1,2-di-O-dodecyl-sn-glycero-3-phosphocholine might be used in small micelles to study the interaction of peptides with model membranes by nuclear magnetic resonance (NMR) experiments.

Biochem/physiol Actions

12:0 Diether PC (phosphocholine) is useful in bicelles formation. Bicelles are suitable to be integrated into standard crystallization protocols and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation. It is found to be a simpler membrane model and is highly compatible with nuclear magnetic resonance (NMR).

Packaging

5 mL Amber Glass Screw Cap Vial (999994P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of a peptide derived from C-terminus of human TRPA1 channel with model membranes mimicking the inner leaflet of the plasma membrane
Witschas K, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(5), 1147-1156 (2015)
Structural and functional roles of ether lipids
Dean JM and Lodhi IJ
Protein & cell, 9(2), 196-206 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service