Skip to Content
Merck
All Photos(1)

Key Documents

860846P

Avanti

CER8(d9)

N-palmitoyl(d9) 6R-hydroxysphingosine, powder

Synonym(s):

13,13,14,14,15,15,16,16,16-nonadeuterio-N-[(E,2S,3R,6R)-1,3,6-trihydroxyoctadec-4-en-2-yl]hexadecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H58D9NO4
CAS Number:
Molecular Weight:
562.96
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 1 mg (860846P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860846P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](NC(CCCCCCCCCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O)([H])[C@]([H])(O)/C=C/[C@@](O)([H])CCCCCCCCCCCC

InChI

1S/C34H67NO4/c1-3-5-7-9-11-13-15-16-17-19-21-23-25-27-34(39)35-32(30-36)33(38)29-28-31(37)26-24-22-20-18-14-12-10-8-6-4-2/h28-29,31-33,36-38H,3-27,30H2,1-2H3,(H,35,39)/b29-28+/t31-,32+,33-/m1/s1/i1D3,3D2,5D2,7D2

InChI key

CAXCJECPKVAJKC-NPZBHXLWSA-N

General description

6-Hydroxysphingosine is one of the moieties that make up the class of ceramides. Ceramides are lipids localized to stratum corneum.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860846P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Principles and Practice of Skin Toxicology, 21-21 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service