Skip to Content
Merck
All Photos(2)

Key Documents

700364O

Avanti

Linoleic acid (d5)

Avanti Research - A Croda Brand

Synonym(s):

(9Z,12Z)-octadeca-9,12-dienoic-17,17,18,18,18-d5 acid, 9,12-Octadecanoic acid (d5), Linolic acid(d5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27D5O2
CAS Number:
Molecular Weight:
285.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

oil

packaging

pkg of 1 × 10 mg (700364O-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Application

Lipidomics

Features and Benefits

Contains 1% BHT

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidic acid induces leaf cell death in Arabidopsis by activating the Rho-related small G protein GTPase-mediated pathway of reactive oxygen species generation
Park J, et al.
Plant Physiology, 134(1), 129-136 (2004)
A hand-held fiber-optic implement for the site-specific delivery of photosensitizer and singlet oxygen
Mahendran A, et al.
Photochemistry and Photobiology, 87(6), 1330-1337 (2011)
Cholesterol induces surface localization of polyphenols in model membranes thus enhancing vesicle stability against lysozyme, but reduces protection of distant double bonds from reactive-oxygen species
Collado A A M, et al.
Biochimica et Biophysica Acta, 1858, 1479-1487 (2016)
Polyunsaturated Fatty Acid Composition of Cerebrospinal Fluid Fractions Shows Their Contribution to Cognitive Resilience of a Pre-symptomatic Alzheimer's Disease Cohort
Fonteh N A, et al.
Frontiers in Physiology, 11, 83-83 (2020)
Christopher M Bobba et al.
Nature communications, 12(1), 289-289 (2021-01-14)
Mechanical ventilation generates injurious forces that exacerbate lung injury. These forces disrupt lung barrier integrity, trigger proinflammatory mediator release, and differentially regulate genes and non-coding oligonucleotides including microRNAs. In this study, we identify miR-146a as a mechanosensitive microRNA in alveolar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service