Key Documents
Z645818
Chemical spill cushions
size small
Synonym(s):
chemical cleanup, spill cleanup, spill safety
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
size
small
description
each cushion absorbs 1.8 Litres
manufacturer/tradename
Clear-spill CCS
availability
not available in ROW, USA and Canada
L × W
23 cm × 23 cm
General description
Cushions can be used with drip pans or in conjection with pads and socks for absorbing larger spills.
Choose from one of the most recent versions:
Certificates of Analysis (COA)