WHA68094002
Whatman® Anotop® 25 Plus syringe filter
pore size 0.02 μm, glass microfiber prefilter
Synonym(s):
Whatman filter, Z747831, syringe filter, syringe filters
About This Item
Recommended Products
material
alumina based membrane
polypropylene housing
sterility
non-sterile
feature
glass microfiber prefilter
packaging
pkg of 50 ea
manufacturer/tradename
Whatman 6809-4002
Whatman Article No. 28420404 (US reference)
diam.
25 mm
volume
≤100 mL
pore size
0.02 μm
fitting
female Luer inlet (lock)
male Luer outlet
Looking for similar products? Visit Product Comparison Guide
Application
- Filtration of tissue culture media
- Clean-up of difficult samples
- Filtration of colloidal material
- Removal of mycoplasma
- HPLC sample preparation
- Biological sample preparation
Features and Benefits
- Capillary pore structure for well-defined particle filtration
- Low protein binding filter for applications requiring high protein recovery
- Integrated glass microfiber prefilter for filtering larger volumes of difficult-to-filter samples
Suitability
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service