SLW1521/16D
Pyrex® Reagent bottles, narrow neck with stopper
capacity 5000 mL
Synonym(s):
Pyrex, SciLabware 1521/16D
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
Pyrex bottle
colorless bottle
glass cap
glass stopper
narrow-mouth bottle
Joint
joint ST/NS neck45/40
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
SciLabware 1521/16D
bottle diam. × H
181 mm × 333 mm , With stopper
capacity
5000 mL
neck I.D.
45 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with ISO 4796-2
- Borosilicate glass with ground neck
- Supplied with solid glass stopper
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service