MT51343010
Mettler Toledo InLab® Easy pH electrode
S7 connector, without cable
All Photos(1)
About This Item
UNSPSC Code:
41115604
NACRES:
NB.79
Recommended Products
material
glass, U membrane
polysulfone shaft
manufacturer/tradename
Mettler-Toledo 51343010
temperature measuring range
0-80 °C
L × W
120 mm × 12 mm
pH
0-14
General description
The Mettler Toledo InLab® Easy are low maintenance, pH combination electrodes with polysulphone shaft, gel electrolyte, ceramic junction and variety of connectors. Ideal for field measurement and educational use
Ideal for field measurement and educational use.
The tough, durable polysulphone shaft with integral bulb protector gives maximum protection against breakage. The use of a gel electrolyte avoids the inconvenience of re-filling the reference electrolyte in the field. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
Ideal for field measurement and educational use.
The tough, durable polysulphone shaft with integral bulb protector gives maximum protection against breakage. The use of a gel electrolyte avoids the inconvenience of re-filling the reference electrolyte in the field. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
- Robust polysulphone shaft
- Low maintenance gel electrolyte
- \Integral bulb protector
- Detachable cable
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.